Anti-ACE2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306147EA 4825 NOK InStock
ORIGTA306147
Anti-ACE2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more