Anti-XIAP Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306170EA 8425 NOK
ORIGTA306170
Anti-XIAP Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR