Anti-BCL2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306171EA 8425 NOK
ORIGTA306171
Anti-BCL2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR