Anti-CASP8 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306206EA 4825 NOK InStock
ORIGTA306206
Anti-CASP8 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: CASP8 (caspase 8, apoptosis-related cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more