Anti-C1QTNF3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306233EA 8425 NOK
ORIGTA306233
Anti-C1QTNF3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: C1QTNF3 (C1q and tumor necrosis factor related protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR