Anti-C1QTNF7 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306282EA 8425 NOK
ORIGTA306282
Anti-C1QTNF7 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: C1QTNF7 (C1q and tumor necrosis factor related protein 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR