Anti-TRPC6 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306349EA 8425 NOK
ORIGTA306349
Anti-TRPC6 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: TRPC6 (transient receptor potential cation channel, subfamily C, member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR