Anti-TNFRSF12A Rabbit Polyclonal Antibody
Leverandør: OriGene
ORIGTA306358EA 8425 NOK
ORIGTA306358
Anti-TNFRSF12A Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: TNFRSF12A (tumor necrosis factor receptor superfamily, member 12A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human