Anti-ORAI2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306419EA 4825 NOK InStock
ORIGTA306419
Anti-ORAI2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-4 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: ORAI2 (ORAI calcium release-activated calcium modulator 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more