Anti-CDNF Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306476EA 8350 NOK
ORIGTA306476
Anti-CDNF Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: CDNF (cerebral dopamine neurotrophic factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR