Anti-SLC9A1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306493EA 8425 NOK
ORIGTA306493
Anti-SLC9A1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: SLC9A1 (solute carrier family 9, subfamily A (NHE1, cation proton antiporter 1), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR