Anti-CAMP Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306515EA 8425 NOK
ORIGTA306515
Anti-CAMP Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Western Blot: 1 μg/ml

Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR