Anti-CXCR4 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306518EA 8425 NOK
ORIGTA306518
Anti-CXCR4 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 5-1 ?g/ml

Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR