Anti-SLITRK2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306523EA 4825 NOK InStock
ORIGTA306523
Anti-SLITRK2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: SLITRK2 (SLIT and NTRK-like family, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more