Anti-SLITRK4 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306528EA 8350 NOK
ORIGTA306528
Anti-SLITRK4 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5 and 1 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: SLITRK4 (SLIT and NTRK-like family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR