Anti-SLITRK6 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306532EA 4825 NOK InStock
ORIGTA306532
Anti-SLITRK6 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Western Blot: 0. 5-1 μg/ml

Type: Primary
Antigen: SLITRK6 (SLIT and NTRK-like family, member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more