Anti-SIRT2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306534EA 4825 NOK InStock
ORIGTA306534
Anti-SIRT2 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions: Western Blot: 1-2 μg/ml

Type: Primary
Antigen: SIRT2 (sirtuin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more