Anti-BAIAP3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306542EA 4825 NOK InStock
ORIGTA306542
Anti-BAIAP3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: BAIAP3 (BAI1-associated protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more