Anti-LGI4 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306544EA 8350 NOK
ORIGTA306544
Anti-LGI4 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: LGI4 (leucine-rich repeat LGI family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR