Anti-NIPSNAP1 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA306548EA 8425 NOK
ORIGTA306548
Anti-NIPSNAP1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: NIPSNAP1 (nipsnap homolog 1 (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR