Anti-GABRA3 Rabbit Antibody

Leverandør: OriGene
ORIGTA309127EA 7575 NOK
ORIGTA309127
Anti-GABRA3 Rabbit Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:100; Immunochemistry: 1:100

Type: Primary
Antigen: GABRA3 (gamma-aminobutyric acid (GABA) A receptor, alpha 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR