Anti-GABRG2 Rabbit Antibody

Leverandør: OriGene
ORIGTA309141EA 7575 NOK
ORIGTA309141
Anti-GABRG2 Rabbit Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GABRG2 (gamma-aminobutyric acid (GABA) A receptor, gamma 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR