Anti-MAP2 Chicken Antibody

Leverandør: OriGene
ORIGTA309162EA 6675 NOK
ORIGTA309162
Anti-MAP2 Chicken Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:20000; Immunofluorescence: 1:2500

Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR