Anti-GRM5 Rabbit Antibody

Leverandør: OriGene
ORIGTA309172EA 7775 NOK
ORIGTA309172
Anti-GRM5 Rabbit Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:500

Type: Primary
Antigen: GRM5 (glutamate receptor, metabotropic 5)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR