Anti-NEFH Mouse Antibody [clone: NAP4]

Leverandør: OriGene
ORIGTA309176EA 7575 NOK
ORIGTA309176
Anti-NEFH Mouse Antibody [clone: NAP4]
Antistoffer
Recommended Dilutions: Western Blot: 1:5000; Immunofluorescence: 1:500

Type: Primary
Antigen: NEFH (neurofilament, heavy polypeptide)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR