Anti-NEFL Mouse Antibody [clone: DA2]

Leverandør: OriGene
ORIGTA309178EA 7575 NOK
ORIGTA309178
Anti-NEFL Mouse Antibody [clone: DA2]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: NEFL (neurofilament, light polypeptide)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR