Anti-NEFM Mouse Antibody [clone: 3H11]

Leverandør: OriGene
ORIGTA309180EA 7575 NOK
ORIGTA309180
Anti-NEFM Mouse Antibody [clone: 3H11]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: NEFM (neurofilament, medium polypeptide)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR