Anti-ENO2 Rabbit Antibody

Leverandør: OriGene
ORIGTA309202EA 6675 NOK
ORIGTA309202
Anti-ENO2 Rabbit Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:500

Type: Primary
Antigen: ENO2 (enolase 2 (gamma, neuronal))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR