Anti-KCNB2 Rabbit Antibody

Leverandør: OriGene
ORIGTA309217EA 7325 NOK
ORIGTA309217
Anti-KCNB2 Rabbit Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: KCNB2 (potassium voltage-gated channel, Shab-related subfamily, member 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR