Anti-SYNJ1 Mouse Antibody [clone: 5H1]

Leverandør: OriGene
ORIGTA309245EA 7325 NOK
ORIGTA309245
Anti-SYNJ1 Mouse Antibody [clone: 5H1]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: SYNJ1 (synaptojanin 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR