Anti-TH Sheep Antibody

Leverandør: OriGene
ORIGTA309267EA 7575 NOK
ORIGTA309267
Anti-TH Sheep Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000; Immunofluorescence: 1:1000

Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR