Anti-UCHL1 Chicken Antibody

Leverandør: OriGene
ORIGTA309272EA 6675 NOK
ORIGTA309272
Anti-UCHL1 Chicken Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:2500; Immunofluorescence: 1:500

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR