Anti-CAMK2A Mouse Monoclonal Antibody [clone: 6G9]

Leverandør: OriGene
ORIGTA309301EA 6800 NOK
ORIGTA309301
Anti-CAMK2A Mouse Monoclonal Antibody [clone: 6G9]
Antistoffer
  • Recommended Dilutions:

Type: Primary
Antigen: CAMK2A (calcium/calmodulin-dependent protein kinase II alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR