Anti-HSPB1 Mouse Monoclonal Antibody [clone: 5D12-A12]

Leverandør: OriGene
ORIGTA309366EA 6300 NOK
ORIGTA309366
Anti-HSPB1 Mouse Monoclonal Antibody [clone: 5D12-A12]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR