Anti-HSPA8 Mouse Monoclonal Antibody [clone: BB70]

Leverandør: OriGene
ORIGTA309384EA 8025 NOK
ORIGTA309384
Anti-HSPA8 Mouse Monoclonal Antibody [clone: BB70]
Antistoffer
  • Recommended Dilutions:

Type: Primary
Antigen: HSPA8 (heat shock 70kDa protein 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR