Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: Hyb-K41220A]

Leverandør: OriGene
ORIGTA309402EA 8025 NOK
ORIGTA309402
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: Hyb-K41220A]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: HSP90AA1 (heat shock protein 90kDa alpha (cytosolic), class A member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR