Anti-KDELR1 Mouse Monoclonal Antibody [clone: KR-10]

Leverandør: OriGene
ORIGTA309411EA 7450 NOK
ORIGTA309411
Anti-KDELR1 Mouse Monoclonal Antibody [clone: KR-10]
Antistoffer
  • Recommended Dilutions:

Type: Primary
Antigen: KDELR1 (KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR