Anti-SLC9A3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309445EA 7875 NOK
ORIGTA309445
Anti-SLC9A3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: SLC9A3 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR