Anti-SH3GL1 Mouse Monoclonal Antibody [clone: S51-1]

Leverandør: OriGene
ORIGTA309473EA 7975 NOK
ORIGTA309473
Anti-SH3GL1 Mouse Monoclonal Antibody [clone: S51-1]
Antistoffer
Recommended Dilutions: Western Blot:1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence:1-10?g/ml

Type: Primary
Antigen: SH3GL1 (SH3-domain GRB2-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR