Anti-slo1 Mouse Monoclonal Antibody [clone: S6-60]

Leverandør: OriGene
ORIGTA309477EA 7975 NOK
ORIGTA309477
Anti-slo1 Mouse Monoclonal Antibody [clone: S6-60]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: slo1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR