Anti-TRPM7 Mouse Monoclonal Antibody [clone: S74-25]

Leverandør: OriGene
ORIGTA309490EA 7975 NOK
ORIGTA309490
Anti-TRPM7 Mouse Monoclonal Antibody [clone: S74-25]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: TRPM7 (transient receptor potential cation channel, subfamily M, member 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR