Anti-SIM1 Goat Polyclonal Antibody

Leverandør: OriGene
ORIGTA309506EA 6325 NOK
ORIGTA309506
Anti-SIM1 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: SIM1 (single-minded family bHLH transcription factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR