Anti-DEFB103A Goat Polyclonal Antibody

Leverandør: OriGene
ORIGTA309519EA 4850 NOK
ORIGTA309519
Anti-DEFB103A Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: DEFB103A (defensin, beta 103A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR