Anti-MTHFR Goat Polyclonal Antibody

Leverandør: OriGene
ORIGTA309530EA 6300 NOK
ORIGTA309530
Anti-MTHFR Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: MTHFR (methylenetetrahydrofolate reductase (NAD(P)H))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR