Anti-FGF5 Goat Polyclonal Antibody

Leverandør: OriGene
ORIGTA309534EA 6300 NOK
ORIGTA309534
Anti-FGF5 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: FGF5 (fibroblast growth factor 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR