Anti-FGFR1 Goat Polyclonal Antibody

Leverandør: OriGene
ORIGTA309547EA 6300 NOK
ORIGTA309547
Anti-FGFR1 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: FGFR1 (fibroblast growth factor receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR