Anti-CDKN2A Goat Polyclonal Antibody

Leverandør: OriGene
ORIGTA309548EA 6300 NOK
ORIGTA309548
Anti-CDKN2A Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR