Anti-AGPAT6 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309568EA 7475 NOK
ORIGTA309568
Anti-AGPAT6 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: AGPAT6 (1-acylglycerol-3-phosphate O-acyltransferase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR