Anti-AQP2 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA309574EA 7850 NOK
ORIGTA309574
Anti-AQP2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: AQP2 (aquaporin 2 (collecting duct))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR