Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]

Leverandør: OriGene
ORIGTA309577EA 9750 NOK
ORIGTA309577
Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]
Antistoffer
Recommended Dilutions:

Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR